Assay Details
Target Gene Details
Entrez Gene ID: | 2634 |
Gene Name: | guanylate binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.1:89106132-89126159 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP2 | NM_004120.4 | 11 | 2615 | NP_004111.2 |
| AI400402.1 | ||||
| AI445567.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546797 | Chr.1:89081456 - 89123873 on Build GRCh38 | Loss |
|
| nsv546796 | Chr.1:89052437 - 89256179 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114437299] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map