Assay Details
Target Gene Details
Entrez Gene ID: | 116238 |
Gene Name: | TLC domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.17:28724348-28727930 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLCD1 | NM_001160407.1 | 4 | 511 | NP_001153879.1 |
| NM_138463.3 | 4 | 644 | NP_612472.1 | |
| XM_006721671.3 | 4 | 447 | XP_006721734.1 | |
| XM_011524278.2 | XP_011522580.1 | |||
| BC014072.2 | 4 | 640 | AAH14072.1 | |
| BX106421.1 | 4 | 512 | ||
| CA453962.1 | 4 | 650 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|

Genomic Map