Assay Details
Target Gene Details
Entrez Gene ID: | 8861 |
Gene Name: | LIM domain binding 1 |
Gene Aliases: |
CLIM-2, CLIM2, LDB-1, NLI |
Location: |
Chr.10:102102100-102120473 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDB1 | NM_001113407.2 | 11 | 1727 | NP_001106878.1 |
| NM_001321612.1 | 11 | 1721 | NP_001308541.1 | |
| NM_003893.4 | 11 | 1900 | NP_003884.1 | |
| XM_017016867.1 | XP_016872356.1 | |||
| XM_017016868.1 | XP_016872357.1 | |||
| XM_017016869.1 | 7 | 1018 | XP_016872358.1 | |
| AB016485.1 | BAA31991.1 | |||
| AF064491.1 | 11 | 1569 | AAC28341.1 | |
| AF068652.1 | 11 | 1540 | AAC77818.1 | |
| BC000482.2 | 11 | 1879 | AAH00482.1 | |
| BC009246.2 | 11 | 1879 | AAH09246.1 |
More Information
Set Membership: |
|

Genomic Map