Assay Details
Target Gene Details
Entrez Gene ID: | 9514 |
Gene Name: | galactose-3-O-sulfotransferase 1 |
Gene Aliases: |
CST |
Location: |
Chr.22:30554635-30574588 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588887 | Chr.22:30557534 - 30599369 on Build GRCh38 | Loss |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80102388] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map