Assay Details
Target Gene Details
Entrez Gene ID: | 11252 |
Gene Name: | protein kinase C and casein kinase substrate in neurons 2 |
Gene Aliases: |
SDPII |
Location: |
Chr.22:42869766-43015690 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PACSIN2 | NM_001184970.1 | 11 | 2016 | NP_001171899.1 |
| NM_001184971.1 | 10 | 1818 | NP_001171900.1 | |
| NM_007229.3 | 11 | 2003 | NP_009160.2 | |
| XM_005261319.3 | 10 | 2405 | XP_005261376.1 | |
| XM_011529846.1 | 12 | 2534 | XP_011528148.1 | |
| XM_011529847.2 | 12 | 2119 | XP_011528149.1 | |
| XM_011529848.2 | 12 | 1971 | XP_011528150.1 | |
| XM_011529849.2 | 12 | 2155 | XP_011528151.1 | |
| XM_011529850.2 | 12 | 2346 | XP_011528152.1 | |
| XM_011529851.2 | 12 | 2036 | XP_011528153.1 | |
| XM_011529852.1 | 11 | 2397 | XP_011528154.1 | |
| XM_011529853.1 | 11 | 2411 | XP_011528155.1 | |
| XM_017028562.1 | 12 | 2000 | XP_016884051.1 | |
| XM_017028563.1 | 12 | 2052 | XP_016884052.1 | |
| XM_017028564.1 | 11 | 2104 | XP_016884053.1 | |
| XM_017028565.1 | 11 | 2016 | XP_016884054.1 | |
| XM_017028566.1 | 11 | 2052 | XP_016884055.1 | |
| XM_017028567.1 | 11 | 2243 | XP_016884056.1 | |
| XM_017028568.1 | 10 | 2274 | XP_016884057.1 | |
| AF128536.1 | 11 | 1989 | AAD41781.1 | |
| AL136845.1 | 10 | 1818 | CAB66779.1 | |
| AL389984.1 | 3 | 656 | CAB97538.1 | |
| AY007086.1 | 2 | 512 | ||
| BC008037.2 | 11 | 2003 | AAH08037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508738 | Chr.22:42839659 - 42917996 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map