Assay Details
Target Gene Details
Entrez Gene ID: | 23568 |
Gene Name: | ADP ribosylation factor like GTPase 2 binding protein |
Gene Aliases: |
BART, BART1, RP66 |
Location: |
Chr.16:57245126-57253635 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL2BP | NM_012106.3 | NP_036238.1 | ||
| XM_011522977.1 | XP_011521279.1 | |||
| AF126062.1 | AAD20633.1 | |||
| AK075050.1 | ||||
| AK298758.1 | ||||
| AK311018.1 | ||||
| BC003087.1 | AAH03087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509621 | Chr.16:57248694 - 57299748 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map