Assay Details
Target Gene Details
Entrez Gene ID: | 957 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 5 |
Gene Aliases: |
CD39L4, NTPDase-5, PCPH |
Location: |
Chr.14:73959420-74019326 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD5 | NM_001249.3 | 16 | 1604 | NP_001240.1 |
| NM_001321984.1 | NP_001308913.1 | |||
| NM_001321985.1 | 17 | 1721 | NP_001308914.1 | |
| NM_001321986.1 | 17 | 1701 | NP_001308915.1 | |
| NM_001321987.1 | 18 | 1848 | NP_001308916.1 | |
| NM_001321988.1 | 16 | 2380 | NP_001308917.1 | |
| XM_005268224.3 | 16 | 1566 | XP_005268281.1 | |
| XM_006720325.3 | XP_006720388.1 | |||
| XM_006720326.2 | 15 | 1466 | XP_006720389.1 | |
| XM_017021813.1 | 17 | 1726 | XP_016877302.1 | |
| XM_017021814.1 | XP_016877303.1 | |||
| XM_017021815.1 | XP_016877304.1 | |||
| XM_017021816.1 | 13 | 1340 | XP_016877305.1 | |
| XM_017021817.1 | XP_016877306.1 | |||
| AF039918.1 | 15 | 1539 | AAC39885.1 | |
| AF136572.1 | 15 | 1575 | AAK82950.1 | |
| AY430094.1 | 15 | 1545 | AAR06666.1 | |
| BC130485.1 | 13 | 1321 | ||
| BC130487.1 | 13 | 1321 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1352 | Chr.14:73951320 - 73996025 on Build GRCh38 | Deletion |
|
| nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain |
|
| dgv1937n100 | Chr.14:73950133 - 74115803 on Build GRCh38 | Gain |
|
| nsv524396 | Chr.14:73958235 - 74023032 on Build GRCh38 | Gain |
|
| nsv1046028 | Chr.14:73957079 - 74032042 on Build GRCh38 | Loss |
|
| nsv1051583 | Chr.14:73953737 - 73995987 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map