Assay Details
Target Gene Details
Entrez Gene ID: | 27295 |
Gene Name: | PDZ and LIM domain 3 |
Gene Aliases: |
ALP |
Location: |
Chr.4:185500660-185535606 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDLIM3 | NM_001114107.4 | 7 | 1252 | NP_001107579.1 |
| NM_001257962.1 | 7 | 1132 | NP_001244891.1 | |
| NM_001257963.1 | 5 | 895 | NP_001244892.1 | |
| NM_014476.5 | 8 | 1396 | NP_055291.2 | |
| NR_047562.1 | 6 | 1064 | ||
| AF002280.1 | 8 | 1318 | AAC16670.1 | |
| AF002282.1 | 7 | 1174 | AAC16672.1 | |
| AF039018.1 | 8 | 1342 | AAB96665.1 | |
| AK129863.1 | 6 | 933 | ||
| AK222966.1 | 8 | 1300 | BAD96686.1 | |
| BC027870.1 | 7 | 1179 | AAH27870.1 | |
| BX647263.1 | 4 | 4773 | ||
| CD625728.1 | ||||
| CR739525.1 | 1 | 353 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv596373 | Chr.4:185451672 - 185605031 on Build GRCh38 | Gain |
|
| dgv5493n100 | Chr.4:185446588 - 185605031 on Build GRCh38 | Gain |
|
| esv3603504 | Chr.4:185498199 - 185517654 on Build GRCh38 | Gain |
|
| esv3603496 | Chr.4:185447616 - 185603060 on Build GRCh38 | Gain |
|
| nsv517035 | Chr.4:185445161 - 185600889 on Build GRCh38 | Gain+Loss |
|
| nsv527026 | Chr.4:185404275 - 185632326 on Build GRCh38 | Gain |
|
| nsv508339 | Chr.4:185501498 - 185533923 on Build GRCh38 | Deletion |
|
| esv2759309 | Chr.4:185391971 - 185553542 on Build GRCh38 | Loss |
|
| nsv1029738 | Chr.4:185450051 - 185515559 on Build GRCh38 | Gain |
|
| nsv4639 | Chr.4:185485731 - 185524173 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138685478] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map