Assay Details
Target Gene Details
Entrez Gene ID: | 3706 |
Gene Name: | inositol-trisphosphate 3-kinase A |
Gene Aliases: |
IP3-3KA, IP3KA |
Location: |
Chr.15:41493858-41503559 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPKA | NM_002220.2 | NP_002211.1 | ||
| XM_011521522.2 | XP_011519824.1 | |||
| BC026331.1 | AAH26331.1 | |||
| X54938.1 | CAA38700.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471241 | Chr.15:41491616 - 41528002 on Build GRCh38 | Loss |
|
| esv33378 | Chr.15:41494307 - 41513562 on Build GRCh38 | Gain |
|
| nsv524476 | Chr.15:41397034 - 41528002 on Build GRCh38 | Loss |
|
| nsv569233 | Chr.15:41492611 - 41513039 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map