Assay Details
Target Gene Details
Entrez Gene ID: | 54943 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C28 |
Gene Aliases: |
C21orf55, C21orf78 |
Location: |
Chr.21:33488055-33491723 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC28 | NM_001040192.2 | 2 | 465 | NP_001035282.1 |
| NM_001320746.1 | 2 | 442 | NP_001307675.1 | |
| NM_017833.4 | 2 | 761 | NP_060303.2 | |
| AF462153.2 | 2 | 751 | AAL59618.2 | |
| AK000468.1 | 2 | 751 | BAA91185.1 | |
| AK225514.1 | 2 | 751 | ||
| DA234338.1 | 2 | 458 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509795 | Chr.21:33457276 - 33521074 on Build GRCh38 | Insertion |
|
| nsv524420 | Chr.21:33455533 - 33491977 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map