Assay Details
Target Gene Details
Entrez Gene ID: | 51115 |
Gene Name: | regulator of microtubule dynamics 1 |
Gene Aliases: |
CGI-90, FAM82B, RMD-1, RMD1 |
Location: |
Chr.8:86467116-86514348 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023510 | Chr.8:86416772 - 86862720 on Build GRCh38 | Loss |
|

Genomic Map