Assay Details
Target Gene Details
Entrez Gene ID: | 84516 |
Gene Name: | dynactin subunit 5 |
Gene Aliases: |
- |
Location: |
Chr.16:23641366-23676207 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCTN5 | NM_001199011.1 | NP_001185940.1 | ||
| NM_001199743.1 | NP_001186672.1 | |||
| NM_032486.3 | NP_115875.1 | |||
| NR_037573.1 | ||||
| DA310703.1 |
Target Gene Details
Entrez Gene ID: | 79728 |
Gene Name: | partner and localizer of BRCA2 |
Gene Aliases: |
FANCN, PNCA3 |
Location: |
Chr.16:23603162-23641357 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PALB2 | NM_024675.3 | NP_078951.2 | ||
| XM_011545946.2 | XP_011544248.1 | |||
| XM_011545948.2 | XP_011544250.1 | |||
| XM_017023672.1 | XP_016879161.1 | |||
| XM_017023673.1 | XP_016879162.1 | |||
| AK097533.1 | BAC05090.1 | |||
| BC044254.2 | AAH44254.1 | |||
| BX647125.1 | ||||
| BX647130.1 | ||||
| CR749637.1 | CAH18431.1 | |||
| DA192828.1 |
More Information
Set Membership: |
|

Genomic Map