Assay Details
Target Gene Details
Entrez Gene ID: | 3818 |
Gene Name: | kallikrein B1 |
Gene Aliases: |
KLK3, PKK, PKKD, PPK |
Location: |
Chr.4:186215714-186258477 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLKB1 | NM_000892.4 | 3 | 248 | NP_000883.2 |
| NM_001318394.1 | 4 | 317 | NP_001305323.1 | |
| NM_001318396.1 | 3 | 248 | NP_001305325.1 | |
| XM_011531930.2 | 3 | 201 | XP_011530232.1 | |
| XM_017008181.1 | 3 | 965 | XP_016863670.1 | |
| XM_017008182.1 | 3 | 201 | XP_016863671.1 | |
| XM_017008183.1 | 3 | 201 | XP_016863672.1 | |
| AK292624.1 | 3 | 249 | ||
| AK297672.1 | 4 | 270 | ||
| AK313378.1 | 3 | 198 | ||
| BC117349.1 | 3 | 160 | ||
| BC117351.1 | 3 | 160 | ||
| CN265612.1 | 4 | 367 | ||
| M13143.2 | 3 | 201 | AAA60153.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1028146 | Chr.4:185582745 - 186417181 on Build GRCh38 | Gain |
|
| nsv830172 | Chr.4:186091611 - 186247270 on Build GRCh38 | Loss |
|
| nsv461864 | Chr.4:186085652 - 186335850 on Build GRCh38 | Gain |
|
| nsv596401 | Chr.4:186151229 - 186445495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map