Assay Details
Target Gene Details
Entrez Gene ID: | 10988 |
Gene Name: | methionyl aminopeptidase 2 |
Gene Aliases: |
MAP2, MNPEP, p67eIF2 |
Location: |
Chr.12:95474046-95515839 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METAP2 | NM_001317182.1 | NP_001304111.1 | ||
| NM_001317183.1 | NP_001304112.1 | |||
| NM_006838.3 | NP_006829.1 | |||
| NR_133673.1 | ||||
| AK125296.1 | 1 | 546 | ||
| AK300836.1 | ||||
| DW409106.1 |
More Information
Set Membership: |
|

Genomic Map