Assay Details
Target Gene Details
Entrez Gene ID: | 388323 |
Gene Name: | glycolipid transfer protein domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.17:4788916-4790589 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLTPD2 | NM_001014985.2 | NP_001014985.2 | ||
| XM_005256626.3 | XP_005256683.1 | |||
| BC150536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833347 | Chr.17:4785297 - 4955981 on Build GRCh38 | Loss |
|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| nsv833346 | Chr.17:4646256 - 4860871 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map