Assay Details
Target Gene Details
Entrez Gene ID: | 10120 |
Gene Name: | ARP1 actin-related protein 1 homolog B, centractin beta |
Gene Aliases: |
ARP1B, CTRN2, PC3 |
Location: |
Chr.2:97655939-97664098 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR1B | NM_005735.3 | NP_005726.1 | ||
| XM_005263854.4 | XP_005263911.1 | |||
| XM_017003116.1 | XP_016858605.1 | |||
| BC004374.2 | AAH04374.1 | |||
| BC006372.2 | AAH06372.1 | |||
| BC010090.2 | AAH10090.1 | |||
| X82207.1 | CAA57691.1 | |||
| X82208.1 | CAA57692.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4026n100 | Chr.2:97360488 - 97665171 on Build GRCh38 | Gain |
|
| esv2763630 | Chr.2:97065396 - 97662587 on Build GRCh38 | Gain+Loss |
|
| dgv4025n100 | Chr.2:97206888 - 97665171 on Build GRCh38 | Gain |
|
| esv3575281 | Chr.2:97235230 - 97660639 on Build GRCh38 | Gain |
|
| nsv582512 | Chr.2:97656171 - 97661341 on Build GRCh38 | Loss |
|
| esv2759077 | Chr.2:96905372 - 97720044 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73959765] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map