Assay Details
Target Gene Details
Entrez Gene ID: | 11189 |
Gene Name: | CUGBP, Elav-like family member 3 |
Gene Aliases: |
BRUNOL1, CAGH4, ERDA4, ETR-1, TNRC4 |
Location: |
Chr.1:151700058-151716821 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELF3 | NM_001172648.3 | 13 | 2622 | NP_001166119.1 |
| NM_001172649.3 | 12 | 2719 | NP_001166120.1 | |
| NM_001291106.1 | 13 | 2866 | NP_001278035.1 | |
| NM_001291107.1 | 13 | 2866 | NP_001278036.1 | |
| NM_007185.6 | 13 | 2869 | NP_009116.3 | |
| XM_006711139.2 | 14 | 2097 | XP_006711202.1 | |
| XM_006711141.2 | 14 | 2094 | XP_006711204.1 | |
| XM_011509111.2 | 12 | 2564 | XP_011507413.1 | |
| XM_011509112.2 | 12 | 1960 | XP_011507414.1 | |
| XM_011509114.2 | 12 | 1928 | XP_011507416.1 | |
| XM_017000145.1 | 12 | 1935 | XP_016855634.1 | |
| XM_017000146.1 | 12 | 1932 | XP_016855635.1 | |
| AB209264.1 | 13 | 2622 | BAD92501.1 | |
| AF284423.1 | AAG14457.1 | |||
| AY165003.1 | 13 | 2859 | AAN73884.1 | |
| BC004145.2 | 9 | 1245 | AAH04145.2 | |
| BC052491.1 | 13 | 2840 | AAH52491.1 | |
| BC068008.1 | 12 | 1896 | AAH68008.1 | |
| U80746.1 | AAB91444.1 | |||
| U80759.1 | AAB91450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952176 | Chr.1:151701725 - 151717424 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map