Assay Details
Target Gene Details
Entrez Gene ID: | 23473 |
Gene Name: | calpain 7 |
Gene Aliases: |
CALPAIN7, PALBH |
Location: |
Chr.3:15206226-15252916 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPN7 | NM_014296.2 | NP_055111.1 | ||
| XM_011533555.2 | XP_011531857.1 | |||
| XM_011533556.2 | XP_011531858.1 | |||
| XM_011533557.2 | XP_011531859.1 | |||
| XM_017006043.1 | XP_016861532.1 | |||
| XM_017006044.1 | XP_016861533.1 | |||
| XM_017006045.1 | XP_016861534.1 | |||
| AB028639.1 | BAA78730.1 | |||
| AK314253.1 | ||||
| BC056202.1 | AAH56202.1 | |||
| BI464220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428413 | Chr.3:15083610 - 15308007 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map