Assay Details
Target Gene Details
Entrez Gene ID: | 22990 |
Gene Name: | pecanex homolog 1 (Drosophila) |
Gene Aliases: |
PCNX, PCNXL1, pecanex |
Location: |
Chr.14:70907405-71115382 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCNX1 | NM_001308160.1 | NP_001295089.1 | ||
| NM_014982.2 | NP_055797.2 | |||
| XM_005267420.4 | XP_005267477.1 | |||
| XM_005267421.4 | XP_005267478.1 | |||
| XM_005267422.4 | XP_005267479.1 | |||
| XM_005267423.4 | XP_005267480.1 | |||
| XM_005267425.4 | XP_005267482.1 | |||
| XM_005267427.4 | XP_005267484.1 | |||
| XM_017021096.1 | XP_016876585.1 | |||
| AB023212.1 | BAA76839.1 | |||
| AF233450.1 | AAK82958.1 | |||
| BC140784.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038007 | Chr.14:70803030 - 71200963 on Build GRCh38 | Gain |
|
| nsv1053303 | Chr.14:70849608 - 70936254 on Build GRCh38 | Gain |
|
| esv3581282 | Chr.14:70904152 - 70913920 on Build GRCh38 | Loss |
|

Genomic Map