Assay Details
Target Gene Details
Entrez Gene ID: | 126626 |
Gene Name: | GA binding protein transcription factor beta subunit 2 |
Gene Aliases: |
GABPB-2 |
Location: |
Chr.1:151070180-151125545 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABPB2 | NM_001323906.1 | 1 | 49 | NP_001310835.1 |
| NM_001323907.1 | 1 | 49 | NP_001310836.1 | |
| NM_001323908.1 | 1 | 49 | NP_001310837.1 | |
| NM_001323909.1 | 1 | 49 | NP_001310838.1 | |
| NM_001323910.1 | 1 | 49 | NP_001310839.1 | |
| NM_001323911.1 | 1 | 49 | NP_001310840.1 | |
| NM_001323912.1 | 1 | 49 | NP_001310841.1 | |
| NM_001323913.1 | 1 | 49 | NP_001310842.1 | |
| NM_144618.2 | NP_653219.1 | |||
| XM_017000246.1 | 1 | 447 | XP_016855735.1 | |
| XM_017000247.1 | 1 | 447 | XP_016855736.1 | |
| BC027033.1 | AAH27033.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map