Assay Details
Target Gene Details
Entrez Gene ID: | 3756 |
Gene Name: | potassium voltage-gated channel subfamily H member 1 |
Gene Aliases: |
EAG, EAG1, Kv10.1, TMBTS, ZLS1, h-eag, hEAG1 |
Location: |
Chr.1:210678314-211134115 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNH1 | NM_002238.3 | 11 | 2445 | NP_002229.1 |
| NM_172362.2 | 11 | 2526 | NP_758872.1 | |
| XM_017001246.1 | 6 | 1664 | XP_016856735.1 | |
| AF078741.1 | 11 | 2311 | AAC68668.1 | |
| AF078742.1 | 11 | 2392 | AAC68669.1 | |
| AJ001366.1 | 11 | 2445 | CAA04700.1 | |
| BC113709.1 | 11 | 2345 | ||
| BC143599.1 | 11 | 2346 | ||
| KT895501.1 | 7 | 1204 | ||
| KT895502.1 | 5 | 751 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549123 | Chr.1:210681473 - 210702850 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map