Assay Details
Target Gene Details
Entrez Gene ID: | 26471 |
Gene Name: | nuclear protein 1, transcriptional regulator |
Gene Aliases: |
COM1, P8 |
Location: |
Chr.16:28537341-28539174 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUPR1 | NM_001042483.1 | NP_001035948.1 | ||
| NM_012385.2 | NP_036517.1 | |||
| AF069073.1 | AAC19384.1 | |||
| AF135266.2 | AAD49221.1 | |||
| BC002434.2 | AAH02434.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509608 | Chr.16:28449797 - 28632895 on Build GRCh38 | Insertion |
|
| esv2758640 | Chr.16:28163817 - 29139232 on Build GRCh38 | Gain+Loss |
|
| nsv1059555 | Chr.16:28373144 - 29195838 on Build GRCh38 | Gain |
|
| nsv833180 | Chr.16:28346157 - 28574165 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map