Assay Details
Target Gene Details
Entrez Gene ID: | 153163 |
Gene Name: | uncharacterized LOC153163 |
Gene Aliases: |
- |
Location: |
Chr.5:122436497-122479087 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGC32805 | NR_051996.1 |
Target Gene Details
Entrez Gene ID: | 9627 |
Gene Name: | synuclein alpha interacting protein |
Gene Aliases: |
SYPH1, Sph1 |
Location: |
Chr.5:122311354-122464219 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 17 - Intron 17 |
More Information
Set Membership: |
|

Genomic Map