Assay Details
Target Gene Details
Entrez Gene ID: | 2005 |
Gene Name: | ELK4, ETS transcription factor |
Gene Aliases: |
SAP1 |
Location: |
Chr.1:205597774-205634077 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELK4 | NM_001973.3 | 5 | 2869 | NP_001964.2 |
| XM_005244950.4 | 6 | 1779 | XP_005245007.1 | |
| XM_005244951.4 | 5 | 2573 | XP_005245008.1 | |
| BC040145.1 | AAH40145.1 | |||
| BE041598.1 | 1 | 134 | ||
| M85165.1 | 5 | 1768 | AAA03631.1 |
More Information
Set Membership: |
|

Genomic Map