Assay Details
Target Gene Details
Entrez Gene ID: | 114905 |
Gene Name: | C1q and tumor necrosis factor related protein 7 |
Gene Aliases: |
CTRP7, ZACRP7 |
Location: |
Chr.4:15339818-15446169 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1QTNF7 | NM_001135170.1 | 1 | 103 | NP_001128642.1 |
| XM_011513772.1 | XP_011512074.1 | |||
| AK314802.1 | 1 | 51 | ||
| BC022187.2 | 1 | 51 | AAH22187.1 | |
| BC024015.2 | 1 | 51 | AAH24015.1 | |
| BX647781.1 | 1 | 103 |
Target Gene Details
Entrez Gene ID: | 101929095 |
Gene Name: | uncharacterized LOC101929095 |
Gene Aliases: |
- |
Location: |
Chr.4:15004942-15427914 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929095 | NR_125911.1 | |||
| DB497821.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114223167] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map