Assay Details
Target Gene Details
Entrez Gene ID: | 9068 |
Gene Name: | angiopoietin like 1 |
Gene Aliases: |
ANG3, ANGPT3, ARP1, AngY, UNQ162, dJ595C2.2 |
Location: |
Chr.1:178849535-178871353 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPTL1 | NM_004673.3 | NP_004664.1 | ||
| XM_005245577.2 | XP_005245634.1 | |||
| AB056476.1 | BAB40691.1 | |||
| AK223467.1 | BAD97187.1 | |||
| AK300218.1 | 1 | 87 | ||
| AY358278.1 | AAQ88645.1 | |||
| BC050640.1 | AAH50640.1 | |||
| BX499002.1 | ||||
| CD356427.1 |
Target Gene Details
Entrez Gene ID: | 55103 |
Gene Name: | Ral GEF with PH domain and SH3 binding motif 2 |
Gene Aliases: |
dJ595C2.1 |
Location: |
Chr.1:178725147-178921842 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RALGPS2 | NM_001286247.1 | NP_001273176.1 | ||
| NM_152663.4 | NP_689876.2 | |||
| XM_006711410.3 | XP_006711473.1 | |||
| XM_006711411.3 | XP_006711474.1 | |||
| XM_011509688.1 | XP_011507990.1 | |||
| XM_017001591.1 | XP_016857080.1 | |||
| AK001106.1 | BAA91506.1 | |||
| AK301753.1 | ||||
| BC047391.1 | AAH47391.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005359 | Chr.1:178848551 - 178898098 on Build GRCh38 | Loss |
|
| nsv525444 | Chr.1:178693726 - 178918428 on Build GRCh38 | Loss |
|
| nsv3710 | Chr.1:178851240 - 178889766 on Build GRCh38 | Insertion |
|
| nsv466206 | Chr.1:178700536 - 178874759 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73039859] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map