Assay Details
Target Gene Details
Entrez Gene ID: | 284382 |
Gene Name: | actin like 9 |
Gene Aliases: |
HSD21 |
Location: |
Chr.19:8697397-8698830 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTL9 | NM_178525.4 | 1 | 1375 | NP_848620.3 |
| AI798166.1 | 1 | 63 | ||
| AK292258.1 | ||||
| AY248901.1 | AAP20052.1 | |||
| BC024028.2 | AAH24028.1 | |||
| BC045752.1 | AAH45752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060576 | Chr.19:8680583 - 8717236 on Build GRCh38 | Loss |
|
| nsv1057425 | Chr.19:8690894 - 8734632 on Build GRCh38 | Gain |
|
| dgv3425n100 | Chr.19:8684212 - 8768019 on Build GRCh38 | Gain |
|
| dgv3426n100 | Chr.19:8685780 - 8717236 on Build GRCh38 | Gain |
|
| esv2762024 | Chr.19:8567703 - 9197018 on Build GRCh38 | Gain |
|
| nsv833735 | Chr.19:8694435 - 8764996 on Build GRCh38 | Gain+Loss |
|
| nsv471705 | Chr.19:8650622 - 8802639 on Build GRCh38 | Gain+Loss |
|
| nsv469747 | Chr.19:8650621 - 8802639 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map