Assay Details
Target Gene Details
Entrez Gene ID: | 102465476 |
Gene Name: | microRNA 6795 |
Gene Aliases: |
hsa-mir-6795 |
Location: |
Chr.19:15179283-15179350 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6795 | NR_106853.1 |
Target Gene Details
Entrez Gene ID: | 4854 |
Gene Name: | notch 3 |
Gene Aliases: |
CADASIL, CADASIL1, CASIL, IMF2, LMNS |
Location: |
Chr.19:15159633-15200981 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOTCH3 | NM_000435.2 | NP_000426.2 | ||
| XM_005259924.4 | XP_005259981.1 | |||
| AB209447.1 | BAD92684.1 | |||
| U97669.1 | AAB91371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3440n100 | Chr.19:15171818 - 15186765 on Build GRCh38 | Gain |
|
| nsv578717 | Chr.19:15159994 - 15241561 on Build GRCh38 | Loss |
|
| dgv3439n100 | Chr.19:15166388 - 15191569 on Build GRCh38 | Gain |
|
| esv34154 | Chr.19:15101623 - 15284812 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map