Assay Details
Target Gene Details
Entrez Gene ID: | 26249 |
Gene Name: | kelch like family member 3 |
Gene Aliases: |
PHA2D |
Location: |
Chr.5:137617500-137736090 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL3 | NM_001257194.1 | 15 | 5510 | NP_001244123.1 |
| NM_001257195.1 | 13 | 5086 | NP_001244124.1 | |
| NM_017415.2 | 15 | 5335 | NP_059111.2 | |
| AB032955.1 | 15 | 5105 | ||
| AF208068.1 | 15 | 5019 | AAF20938.1 | |
| AF208069.1 | 13 | 5123 | AAF20939.1 | |
| AF208070.1 | 15 | 5510 | AAF20995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830499 | Chr.5:137524807 - 137685237 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map