Assay Details
Target Gene Details
Entrez Gene ID: | 2563 |
Gene Name: | gamma-aminobutyric acid type A receptor delta subunit |
Gene Aliases: |
EIG10, EJM7, GEFSP5 |
Location: |
Chr.1:2019329-2030753 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABRD | NM_000815.4 | 9 | 1458 | NP_000806.2 |
| XM_011541194.2 | 9 | 3370 | XP_011539496.1 | |
| XM_017000936.1 | 8 | 4504 | XP_016856425.1 | |
| AF016917.1 | AAB70007.1 | |||
| AI828107.1 | 1 | 464 | ||
| AI936907.1 | 1 | 464 | ||
| AK290861.1 | 9 | 1439 | ||
| BC033801.1 | 9 | 1374 | AAH33801.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577697 | Chr.1:2028174 - 2045349 on Build GRCh38 | Loss |
|
| nsv951425 | Chr.1:2018162 - 2112261 on Build GRCh38 | Deletion |
|
| nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss |
|
| nsv834446 | Chr.1:1960604 - 2156435 on Build GRCh38 | Loss |
|
| esv2758913 | Chr.1:1389149 - 2092899 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map