Assay Details
Target Gene Details
Entrez Gene ID: | 79137 |
Gene Name: | family with sequence similarity 134 member A |
Gene Aliases: |
C2orf17, MAG-2 |
Location: |
Chr.2:219178190-219185475 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM134A | NM_001321109.1 | NP_001308038.1 | ||
| NM_001321110.1 | NP_001308039.1 | |||
| NM_024293.5 | NP_077269.3 | |||
| XM_005246848.2 | XP_005246905.1 | |||
| AK074983.1 | BAC11332.1 | |||
| AK091011.1 | ||||
| BG700888.1 | ||||
| DA172039.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3170 | Chr.2:219173471 - 219201899 on Build GRCh38 | Deletion |
|
| nsv508200 | Chr.2:219134744 - 219202175 on Build GRCh38 | Deletion |
|
| esv3893555 | Chr.2:219172722 - 219273269 on Build GRCh38 | Loss |
|

Genomic Map