Assay Details
Target Gene Details
Entrez Gene ID: | 10293 |
Gene Name: | TRAF interacting protein |
Gene Aliases: |
RNF206, SCKL9, TRIP |
Location: |
Chr.3:49828595-49856584 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAIP | NM_005879.2 | NP_005870.2 | ||
| XM_011533264.1 | XP_011531566.1 | |||
| XM_017005526.1 | XP_016861015.1 | |||
| AB451320.1 | ||||
| AB451449.1 | ||||
| AK225873.1 | ||||
| AK292172.1 | ||||
| BC000310.2 | AAH00310.1 | |||
| BC019283.1 | AAH19283.1 | |||
| U77845.1 | AAB52993.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834688 | Chr.3:49773682 - 49981866 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map