Assay Details
Target Gene Details
Entrez Gene ID: | 85440 |
Gene Name: | dedicator of cytokinesis 7 |
Gene Aliases: |
EIEE23, ZIR2 |
Location: |
Chr.1:62454726-62688368 on Build GRCh38 |
Assay Gene Location: | Within Exon 52 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK7 | NM_001271999.1 | 48 | 6396 | NP_001258928.1 |
| NM_001272000.1 | 48 | 6309 | NP_001258929.1 | |
| NM_001272001.1 | 47 | 6303 | NP_001258930.1 | |
| NM_033407.3 | 48 | 6336 | NP_212132.2 | |
| XM_005271292.2 | 49 | 6392 | XP_005271349.1 | |
| XM_011542326.2 | 49 | 6419 | XP_011540628.1 | |
| XM_011542327.2 | 48 | 6413 | XP_011540629.1 | |
| XM_011542328.2 | 48 | 6404 | XP_011540630.1 | |
| XM_017002639.1 | 47 | 6320 | XP_016858128.1 | |
| AB051558.1 | 29 | 3812 | BAB21862.1 | |
| AK055296.1 | 1 | 1607 | ||
| AK055493.1 | 15 | 1953 | BAB70933.1 | |
| AK055905.1 | 20 | 2828 | BAB71042.1 | |
| AK091479.1 | 22 | 2803 | ||
| AK125049.1 | 34 | 4481 | BAC86032.1 | |
| AK299737.1 | 20 | 2519 | ||
| BC016392.1 | 17 | 2300 | ||
| BC112387.1 | 8 | 1055 | ||
| DQ118679.1 | 45 | 5881 | AAZ38451.1 | |
| DQ118680.1 | 44 | 5875 | AAZ38452.1 | |
| DQ341187.1 | 48 | 6303 |
More Information
Set Membership: |
|

Genomic Map