Assay Details
Target Gene Details
Entrez Gene ID: | 27145 |
Gene Name: | filamin A interacting protein 1 |
Gene Aliases: |
FILIP |
Location: |
Chr.6:75294895-75493829 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FILIP1 | NM_001289987.1 | 3 | 809 | NP_001276916.1 |
| NM_001300866.1 | 2 | 643 | NP_001287795.1 | |
| NM_015687.3 | 2 | 643 | NP_056502.1 | |
| XM_005248713.3 | 2 | 614 | XP_005248770.1 | |
| XM_005248715.4 | 2 | 614 | XP_005248772.1 | |
| AB086011.1 | 1 | 227 | BAC77067.1 | |
| AK097021.1 | 2 | 483 | BAC04928.1 | |
| AK290232.1 | 2 | 355 | ||
| AL832009.2 | 2 | 443 | CAD89912.1 | |
| BC136443.1 | 2 | 398 | ||
| BC136444.1 | 3 | 564 | ||
| BX647178.1 | 2 | 594 |
Target Gene Details
Entrez Gene ID: | 101928540 |
Gene Name: | uncharacterized LOC101928540 |
Gene Aliases: |
- |
Location: |
Chr.6:75383186-75458900 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928540 | NR_125859.1 | |||
| DB456186.1 | ||||
| HY214668.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890880 | Chr.6:75364726 - 75498207 on Build GRCh38 | Loss |
|

Genomic Map