Assay Details
Target Gene Details
Entrez Gene ID: | 55531 |
Gene Name: | ELMO domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.11:107591091-107666779 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMOD1 | NM_001130037.1 | NP_001123509.1 | ||
| NM_001308018.1 | NP_001294947.1 | |||
| NM_018712.3 | NP_061182.3 | |||
| XM_017017994.1 | XP_016873483.1 | |||
| XM_017017995.1 | XP_016873484.1 | |||
| AK054794.1 | ||||
| AK297351.1 | ||||
| AK303689.1 | ||||
| BC028725.2 |
Target Gene Details
Entrez Gene ID: | 643923 |
Gene Name: | uncharacterized LOC643923 |
Gene Aliases: |
- |
Location: |
Chr.11:107591745-107593223 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC643923 | NR_028328.1 | |||
| AK058066.1 | BAB71648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556376 | Chr.11:107380125 - 107615872 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map