Assay Details
Target Gene Details
Entrez Gene ID: | 145694 |
Gene Name: | uncharacterized LOC145694 |
Gene Aliases: |
- |
Location: |
Chr.15:69396904-69415029 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC145694 | NR_132969.1 | |||
| NR_132970.1 | ||||
| NR_132971.1 | ||||
| AK056793.1 | ||||
| CN360125.1 | ||||
| DB499817.1 |
Target Gene Details
Entrez Gene ID: | 54852 |
Gene Name: | progestin and adipoQ receptor family member 5 |
Gene Aliases: |
MPRG |
Location: |
Chr.15:69284134-69407637 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAQR5 | NM_001104554.1 | 9 | 2851 | NP_001098024.1 |
| NM_017705.3 | 9 | 2561 | NP_060175.3 | |
| XM_011521723.2 | 8 | 2440 | XP_011520025.1 | |
| XM_011521725.2 | 7 | 2259 | XP_011520027.1 | |
| XM_017022368.1 | 6 | 2117 | XP_016877857.1 | |
| BC039234.1 | 10 | 2561 | AAH39234.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760037 | Chr.15:69342413 - 69499860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map