Assay Details
Target Gene Details
Entrez Gene ID: | 55863 |
Gene Name: | transmembrane protein 126B |
Gene Aliases: |
HT007 |
Location: |
Chr.11:85628573-85636539 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM126B | NM_001193537.2 | NP_001180466.1 | ||
| NM_001193538.2 | NP_001180467.1 | |||
| NM_001256546.1 | NP_001243475.1 | |||
| NM_001256547.1 | NP_001243476.1 | |||
| NM_018480.4 | NP_060950.3 | |||
| XM_011545165.2 | XP_011543467.1 | |||
| AF220193.1 | 3 | 277 | AAF67658.1 | |
| AF246240.1 | AAG44487.1 | |||
| AK291150.1 | ||||
| AK298189.1 | ||||
| AV728059.1 | ||||
| BC012065.1 | AAH12065.1 | |||
| BC017574.1 | AAH17574.1 | |||
| BC038933.1 | AAH38933.1 | |||
| BC107900.1 | AAI07901.1 | |||
| BE781669.1 | ||||
| BF244908.1 | ||||
| BG170232.1 | ||||
| BG623309.1 | ||||
| BG776073.1 | ||||
| BM746060.1 | ||||
| BP264186.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759845 | Chr.11:85469436 - 85892945 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map