Assay Details
Target Gene Details
Entrez Gene ID: | 116512 |
Gene Name: | MAS related GPR family member D |
Gene Aliases: |
MRGD, TGR7 |
Location: |
Chr.11:68980021-68980986 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRGPRD | NM_198923.2 | 1 | 774 | NP_944605.2 |
| AB154410.1 | 1 | 774 | BAD20638.1 | |
| AY427820.1 | 1 | 774 | AAR05120.1 |
More Information
Set Membership: |
|

Genomic Map