Assay Details
Target Gene Details
Entrez Gene ID: | 4839 |
Gene Name: | NOP2 nucleolar protein |
Gene Aliases: |
NOL1, NOP120, NSUN1, p120 |
Location: |
Chr.12:6556870-6568332 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOP2 | NM_001033714.2 | NP_001028886.1 | ||
| NM_001258308.1 | NP_001245237.1 | |||
| NM_001258309.1 | NP_001245238.1 | |||
| NM_001258310.1 | NP_001245239.1 | |||
| NM_006170.3 | NP_006161.2 | |||
| XM_005253691.1 | XP_005253748.1 | |||
| AK056208.1 | ||||
| AK129852.1 | ||||
| AK307582.1 | ||||
| BC000656.2 | AAH00656.1 | |||
| BC065257.1 | ||||
| BC082985.1 | ||||
| BC106072.1 | AAI06073.1 | |||
| BC128183.1 | ||||
| BC128184.1 | ||||
| M32110.1 | AAA36398.1 | |||
| X55504.1 | CAA39119.1 |
More Information
Set Membership: |
|

Genomic Map