Assay Details
Target Gene Details
Entrez Gene ID: | 57565 |
Gene Name: | kelch like family member 14 |
Gene Aliases: |
- |
Location: |
Chr.18:32672673-32773011 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL14 | NM_020805.2 | 9 | 3445 | NP_065856.1 |
| AB037805.1 | 9 | 3445 |
More Information
Set Membership: |
|

Genomic Map