Assay Details
Target Gene Details
Entrez Gene ID: | 8811 |
Gene Name: | galanin receptor 2 |
Gene Aliases: |
GAL2-R, GALNR2, GALR-2 |
Location: |
Chr.17:76070822-76077541 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALR2 | NM_003857.3 | 2 | 923 | NP_003848.1 |
| XM_011525427.2 | 4 | 2080 | XP_011523729.1 | |
| AF040630.1 | 2 | 952 | AAC39634.1 | |
| AF080586.1 | 2 | 865 | AAD08671.1 | |
| BC069130.1 | 2 | 921 | AAH69130.1 | |
| BC074914.2 | 2 | 887 | AAH74914.1 | |
| BC074915.2 | 2 | 887 | AAH74915.1 | |
| BC109051.1 | 2 | 890 | AAI09052.1 | |
| BC109052.1 | 2 | 889 | AAI09053.1 | |
| BU631750.1 | 1 | 448 | ||
| EF577401.1 | 2 | 840 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2142 | Chr.17:76070185 - 76087802 on Build GRCh38 | Insertion |
|
| nsv833546 | Chr.17:76003077 - 76186972 on Build GRCh38 | Loss |
|
| nsv509674 | Chr.17:76052197 - 76106436 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map