Assay Details
Target Gene Details
Entrez Gene ID: | 90806 |
Gene Name: | angel homolog 2 |
Gene Aliases: |
Ccr4d, KIAA0759L |
Location: |
Chr.1:212992182-213015875 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
More Information
Set Membership: |
|

Genomic Map