Assay Details
Target Gene Details
Entrez Gene ID: | 57623 |
Gene Name: | zinc finger and AT-hook domain containing |
Gene Aliases: |
AITD3, ZFAT1, ZNF406 |
Location: |
Chr.8:134477788-134713049 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFAT | NM_001029939.3 | 17 | 4398 | NP_001025110.2 |
| NM_001167583.2 | 16 | 4239 | NP_001161055.1 | |
| NM_001174157.1 | 15 | 4130 | NP_001167628.1 | |
| NM_001174158.1 | 14 | 3981 | NP_001167629.1 | |
| NM_001289394.1 | 17 | 4475 | NP_001276323.1 | |
| NM_020863.3 | 16 | 4316 | NP_065914.2 | |
| NR_110323.1 | 17 | 4482 | ||
| XM_011517203.1 | 16 | 4213 | XP_011515505.1 | |
| XM_011517204.2 | 15 | 4036 | XP_011515506.1 | |
| AB040918.1 | 14 | 3714 | BAA96009.1 | |
| AB167738.1 | 16 | 4305 | BAD12567.1 | |
| AB167739.1 | 17 | 4464 | BAD12568.1 | |
| AB167740.1 | 17 | 4391 | BAD12569.1 | |
| AK094089.1 | 6 | 2169 | ||
| AK300324.1 | 16 | 4235 | ||
| AK301413.1 | 6 | 1494 | ||
| AL832066.1 | 17 | 4482 | ||
| BC012989.2 | 14 | 3972 | AAH12989.1 | |
| BC025423.2 | AAH25423.1 | |||
| BC046180.1 | 6 | 1240 | AAH46180.1 | |
| BC098405.1 | 16 | 4129 | AAH98405.1 | |
| BI964061.1 | 1 | 387 | ||
| BM685256.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660261 | Chr.8:133867502 - 134652395 on Build GRCh38 | Deletion |
|
| esv25822 | Chr.8:134461813 - 134489442 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map