Assay Details
Target Gene Details
Entrez Gene ID: | 7421 |
Gene Name: | vitamin D (1,25- dihydroxyvitamin D3) receptor |
Gene Aliases: |
NR1I1, PPP1R163 |
Location: |
Chr.12:47841537-47905031 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VDR | NM_000376.2 | 10 | 2361 | NP_000367.1 |
| NM_001017535.1 | 11 | 2483 | NP_001017535.1 | |
| NM_001017536.1 | 10 | 2752 | NP_001017536.1 | |
| AK024830.1 | 1 | 485 | ||
| AK225844.1 | 10 | 2360 | ||
| BC060832.1 | 10 | 2724 | AAH60832.1 | |
| J03258.1 | 10 | 2316 | AAA61273.1 |
More Information
Set Membership: |
|

Genomic Map