Assay Details
Target Gene Details
Entrez Gene ID: | 144402 |
Gene Name: | copine 8 |
Gene Aliases: |
- |
Location: |
Chr.12:38652200-38908593 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPNE8 | NM_153634.2 | NP_705898.1 | ||
| XM_017018851.1 | XP_016874340.1 | |||
| XM_017018852.1 | XP_016874341.1 | |||
| AK074223.1 | ||||
| AK094998.1 | ||||
| AK098593.1 | BAC05344.1 | |||
| AK307908.1 | ||||
| AY177785.1 | AAO21123.1 | |||
| BC046366.1 | AAH46366.1 | |||
| BC110577.1 | AAI10578.1 | |||
| BC110578.1 | AAI10579.1 | |||
| BX538055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv558574 | Chr.12:37848227 - 38793606 on Build GRCh38 | Gain |
|
| esv2761088 | Chr.12:37463949 - 38827290 on Build GRCh38 | Gain |
|
| nsv558350 | Chr.12:37533312 - 39041381 on Build GRCh38 | Gain |
|
| nsv1045040 | Chr.12:38644312 - 38690292 on Build GRCh38 | Gain |
|
| dgv1488n100 | Chr.12:38565843 - 38729051 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map