Assay Details
Target Gene Details
Entrez Gene ID: | 6821 |
Gene Name: | sulfite oxidase |
Gene Aliases: |
- |
Location: |
Chr.12:55996776-56005525 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUOX | NM_000456.2 | 6 | 774 | NP_000447.2 |
| NM_001032386.1 | 5 | 648 | NP_001027558.1 | |
| NM_001032387.1 | 4 | 539 | NP_001027559.1 | |
| XM_005269112.1 | 5 | 630 | XP_005269169.1 | |
| XM_017019905.1 | 7 | 1320 | XP_016875394.1 | |
| XM_017019906.1 | 5 | 608 | XP_016875395.1 | |
| XM_017019907.1 | 6 | 1389 | XP_016875396.1 | |
| XM_017019908.1 | 4 | 549 | XP_016875397.1 | |
| AK312763.1 | 4 | 513 | ||
| AU121557.1 | 5 | 620 | ||
| AU126450.1 | ||||
| BC065193.1 | 4 | 570 | ||
| HM013717.1 | 3 | 457 | ||
| L31573.1 | 3 | 1180 | AAA74886.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832424 | Chr.12:55881442 - 56073133 on Build GRCh38 | Loss |
|
| nsv1051961 | Chr.12:55868436 - 56235643 on Build GRCh38 | Gain |
|
| nsv832427 | Chr.12:55998761 - 56118990 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map