Assay Details
Target Gene Details
Entrez Gene ID: | 4907 |
Gene Name: | 5'-nucleotidase ecto |
Gene Aliases: |
CALJA, CD73, E5NT, NT, NT5, NTE, eN, eNT |
Location: |
Chr.6:85449584-85495791 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5E | NM_001204813.1 | 2 | 945 | NP_001191742.1 |
| NM_002526.3 | 2 | 945 | NP_002517.1 | |
| AK075008.1 | 2 | 420 | ||
| AK314661.1 | 2 | 459 | ||
| BC015940.1 | 2 | 437 | AAH15940.1 | |
| BC065937.1 | 2 | 414 | AAH65937.1 | |
| DQ186653.1 | 2 | 408 | ABA39834.1 | |
| X55740.1 | 2 | 438 | CAA39271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604092 | Chr.6:85405048 - 85554656 on Build GRCh38 | Loss |
|
| nsv520112 | Chr.6:85435157 - 85538041 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map