Assay Details
Target Gene Details
Entrez Gene ID: | 84101 |
Gene Name: | ubiquitin specific peptidase 44 |
Gene Aliases: |
- |
Location: |
Chr.12:95516560-95551529 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP44 | NM_001042403.2 | 6 | 2255 | NP_001035862.1 |
| NM_001278393.1 | 6 | 2471 | NP_001265322.1 | |
| NM_032147.4 | 6 | 2400 | NP_115523.2 | |
| NR_125360.1 | 4 | 1985 | ||
| XM_005269172.1 | 7 | 2487 | XP_005269229.1 | |
| XM_005269173.1 | 7 | 2564 | XP_005269230.1 | |
| XM_005269174.1 | 7 | 2331 | XP_005269231.1 | |
| XM_011538800.2 | 8 | 2514 | XP_011537102.1 | |
| XM_011538801.1 | 8 | 2374 | XP_011537103.1 | |
| XM_011538802.1 | 7 | 2334 | XP_011537104.1 | |
| XM_011538803.1 | 7 | 2339 | XP_011537105.1 | |
| XM_011538804.2 | 7 | 2547 | XP_011537106.1 | |
| XM_011538805.1 | 7 | 3188 | XP_011537107.1 | |
| XM_017020009.1 | 6 | 2389 | XP_016875498.1 | |
| XM_017020010.1 | 6 | 2241 | XP_016875499.1 | |
| XM_017020011.1 | 7 | 2267 | XP_016875500.1 | |
| AK027434.1 | 6 | 2205 | ||
| AK315697.1 | ||||
| AL136825.1 | 6 | 2361 | CAB66759.1 | |
| BC030704.1 | 6 | 2236 | AAH30704.1 | |
| HY021046.1 |
More Information
Set Membership: |
|

Genomic Map