Assay Details
Target Gene Details
Entrez Gene ID: | 286204 |
Gene Name: | crumbs 2, cell polarity complex component |
Gene Aliases: |
FSGS9, VMCKD |
Location: |
Chr.9:123354065-123380335 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRB2 | NM_173689.6 | 7 | 1750 | NP_775960.4 |
| NR_104603.1 | 2 | 770 | ||
| XM_005251934.2 | 2 | 770 | XP_005251991.1 | |
| XM_011518556.2 | 7 | 1750 | XP_011516858.1 | |
| XM_011518557.2 | 7 | 1752 | XP_011516859.1 | |
| XM_011518558.2 | 8 | 2112 | XP_011516860.1 | |
| AK123000.1 | 7 | 1740 | ||
| AK126775.1 | 2 | 770 | BAC86684.1 | |
| AY720432.1 | 7 | 1656 | AAU14134.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615285 | Chr.9:123365719 - 123425419 on Build GRCh38 | Loss |
|
| nsv518647 | Chr.9:123287696 - 123433925 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map