Assay Details
Target Gene Details
Entrez Gene ID: | 79414 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 3 |
Gene Aliases: |
FIGLER1, SALM4 |
Location: |
Chr.19:35937120-35945195 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRFN3 | NM_024509.1 | 3 | 2414 | NP_078785.1 |
| XM_017027302.1 | 2 | 1979 | XP_016882791.1 | |
| AK172754.1 | 2 | 1962 | BAD18740.1 | |
| AY358127.1 | 3 | 2694 | AAQ88494.1 | |
| BC003578.1 | 3 | 2414 | AAH03578.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv51n68 | Chr.19:35841011 - 36016692 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map